Learn More
Brady™ All-weather Vinyl Labels
Product Code. 17831789
Change view
Click to view available options
Dimensions (LxW):
38.10 mm (W) x 25.40 mm (H)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) unitSize
17831789 38.10 mm (W) x 25.40 mm (H) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17831789 Supplier Brady™ Supplier No. 176807
Only null left
Add to basket
Add to basket
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.
  • Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.
  • Versatile: Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Tough: Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Adhesive: Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Long-lasting: Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Certifications/Compliance BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Gloss
Format Die-cut
Material Vinyl
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling;Electrical labelling;Facility identification;Lean and 5S labelling;Lockout tagout;Security identification;Pipe and valve marking
For Use With (Equipment) BMP51;M511;M510
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model M5-31-595
No. per Pack 250 Label(s) / Box
Quantity 1
Size 38.10 mm (W) x 25.40 mm (H)
Temperature Range (English) -40°F to 180°F
Temperature Range (Metric) -40°C to 82°C
Thickness (English) 0.004 in
Thickness (Metric) 0.102 mm
Dimensions (LxW) 38.10 mm (W) x 25.40 mm (H)
Length (English) 1 in.
Length (Metric) 25.40 mm
Width (English) 1.5 in
Width (Metric) 38.10 mm
Width (English) Printable Area 38.10 mm
Height (English) 1 in
Height (Metric) 25.40 mm
Alternate Part Number M5-31-595