Learn More
Brady™ All-weather Vinyl Labels
Product Code. 17970700
Change view
Click to view available options
Dimensions (L x W):
12.70 mm (W) x 6.10 m (L)
12.70 mm (W) x 7.62 m (L)
19.05 mm (W) x 6.10 m (L)
19.05 mm (W) x 7.62 m (L)
25.40 mm (W) x 6.10 m (L)
25.40 mm (W) x 7.62 m (L)
38.10 mm (W) x 6.10 m (L)
38.10 mm (W) x 7.62 m (L)
6.35 mm (W) x 7.62 m (L)
9.53 mm (W) x 7.62 m (L)
Color:
Black, White
Blue, White
Green, White
Orange
Red, White
Transparent, Clear
Transparent, Clear, White
White
White, Red
Yellow
For Use With (Equipment):
BMP41, BMP51, BMP53, M511, M410, M510
BMP51, BMP53, M511, M510
Unit Size:
Each
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color For Use With (Equipment) unitSize
17970700 12.70 mm (W) x 6.10 m (L) Transparent, Clear, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17881809 6.35 mm (W) x 7.62 m (L) White BMP41, BMP51, BMP53, M511, M410, M510 Each
17851819 6.35 mm (W) x 7.62 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510 Each
17941940 9.53 mm (W) x 7.62 m (L) Red, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17910770 9.53 mm (W) x 7.62 m (L) White BMP41, BMP51, BMP53, M511, M410, M510 Each
17940710 9.53 mm (W) x 7.62 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510 Each
17961960 12.70 mm (W) x 7.62 m (L) Black, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17910700 12.70 mm (W) x 7.62 m (L) Blue, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17900700 12.70 mm (W) x 6.10 m (L) Transparent, Clear BMP41, BMP51, BMP53, M511, M410, M510 Each
17920670 12.70 mm (W) x 7.62 m (L) Green, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17970760 12.70 mm (W) x 7.62 m (L) Orange BMP41, BMP51, BMP53, M511, M410, M510 Each
17930700 12.70 mm (W) x 7.62 m (L) Red, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17950700 12.70 mm (W) x 7.62 m (L) White BMP41, BMP51, BMP53, M511, M410, M510 Each
17920700 12.70 mm (W) x 7.62 m (L) White, Red BMP41, BMP51, BMP53, M511, M410, M510 Each
17940670 12.70 mm (W) x 7.62 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510 Each
17941960 19.05 mm (W) x 7.62 m (L) Black, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17921960 19.05 mm (W) x 7.62 m (L) Blue, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17901940 19.05 mm (W) x 6.10 m (L) Transparent, Clear BMP41, BMP51, BMP53, M511, M410, M510 Each
17971960 19.05 mm (W) x 6.10 m (L) Transparent, Clear, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17931950 19.05 mm (W) x 7.62 m (L) Green, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17921950 19.05 mm (W) x 7.62 m (L) Orange BMP41, BMP51, BMP53, M511, M410, M510 Each
17941950 19.05 mm (W) x 7.62 m (L) Red, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17981950 19.05 mm (W) x 7.62 m (L) White BMP41, BMP51, BMP53, M511, M410, M510 Each
17951950 19.05 mm (W) x 7.62 m (L) White, Red BMP41, BMP51, BMP53, M511, M410, M510 Each
17951960 19.05 mm (W) x 7.62 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510 Each
17960670 25.40 mm (W) x 7.62 m (L) Black, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17930670 25.40 mm (W) x 7.62 m (L) Blue, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17910690 25.40 mm (W) x 6.10 m (L) Transparent, Clear BMP41, BMP51, BMP53, M511, M410, M510 Each
17980700 25.40 mm (W) x 6.10 m (L) Transparent, Clear, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17960690 25.40 mm (W) x 7.62 m (L) Green, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17960700 25.40 mm (W) x 7.62 m (L) Orange BMP41, BMP51, BMP53, M511, M410, M510 Each
17930690 25.40 mm (W) x 7.62 m (L) Red, White BMP41, BMP51, BMP53, M511, M410, M510 Each
17970690 25.40 mm (W) x 7.62 m (L) White BMP41, BMP51, BMP53, M511, M410, M510 Each
17950690 25.40 mm (W) x 7.62 m (L) White, Red BMP41, BMP51, BMP53, M511, M410, M510 Each
17940690 25.40 mm (W) x 7.62 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510 Each
17891809 38.10 mm (W) x 7.62 m (L) Black, White BMP51, BMP53, M511, M510 Each
17811789 38.10 mm (W) x 7.62 m (L) Blue, White BMP51, BMP53, M511, M510 Each
17891709 38.10 mm (W) x 6.10 m (L) Transparent, Clear BMP51, BMP53, M511, M510 Each
17801819 38.10 mm (W) x 6.10 m (L) Transparent, Clear, White BMP51, BMP53, M511, M510 Each
17821769 38.10 mm (W) x 7.62 m (L) Green, White BMP51, BMP53, M511, M510 Each
17871819 38.10 mm (W) x 7.62 m (L) Orange BMP51, BMP53, M511, M510 Each
17811769 38.10 mm (W) x 7.62 m (L) Red, White BMP51, BMP53, M511, M510 Each
17821719 38.10 mm (W) x 7.62 m (L) White BMP51, BMP53, M511, M510 Each
17861819 38.10 mm (W) x 7.62 m (L) White, Red BMP51, BMP53, M511, M510 Each
17861829 38.10 mm (W) x 7.62 m (L) Yellow BMP51, BMP53, M511, M510 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
Product Code. 17970700 Supplier Brady™ Supplier No. 170822
Only null left
Add to basket
Add to basket
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
  • Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Permanent Acrylic
Coating None
Color Transparent, Clear, White
Finish Gloss
Format Continuous
Material Vinyl
Printable Sides Top
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking
For Use With (Equipment) BMP41, BMP51, BMP53, M511, M410, M510
Min. Operating Temperature (Metric) -40°C
Model M4C-500-595-CL-WT
No. per Pack 1 Label(s) / Box
Quantity 1
Size 12.70 mm (W) x 6.10 m (L)
Temperature Range (English) -40°F to 180°F
Temperature Range (Metric) -40°C to 82°C
Thickness (English) 0.004 in
Thickness (Metric) 0.102 mm
Dimensions (L x W) 12.70 mm (W) x 6.10 m (L)
Length (English) 20 ft.
Length (Metric) 6.10 m
Width (English) 0.5 in
Width (Metric) 12.70 mm
Width (English) Printable Area 12.70 mm
Height (English) Printable Area 0.5 in.
Alternate Part Number M4C-500-595-CL-WT