Learn More
R&D Systems™ Alkaline Phosphatase, Tissue Non-Specific Assay Kit (Luminescent)
Shop All Bio Techne ProductsSpecifications
Specifications
| Gene | Alkaline Phosphatase, Tissue Non-Specific |
| Gene ID (Entrez) | 249 |
| Gene Alias | Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Product Line | Novus |
| Quantity | 1 Kit |