missing translation for 'onlineSavingsMsg'
Learn More

Alizarin, 94%

Product Code. 11407027
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11407027

Brand: Thermo Scientific Alfa Aesar A14404.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol

Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

Solubility
Soluble in hexane and chloroform. Slightly soluble in water.

Notes
Incompatible with alkalis and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001201
InChI Key RGCKGOZRHPZPFP-UHFFFAOYSA-N
Synonym alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
PubChem CID 6293
ChEBI CHEBI:16866
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O

Specifications

Melting Point 286°C to 290°C
Boiling Point ∼430°C (sublimation)
Quantity 500 g
Beilstein 1914037
Merck Index 14,251
Solubility Information Soluble in hexane and chloroform. Slightly soluble in water.
Formula Weight 240.21
Percent Purity 94%
Chemical Name or Material Alizarin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.