Learn More
Deltalab™ Calibrated Inoculating Loops
Product Code. 15336045
Product Code. 15336045
Brand: Deltalab™ 302753
Only null left
Add to basket
Add to basket
Use both qualitative and quantitative techniques with Deltalab™ Calibrated Inoculating Loops, Double use loops feature a loop for streaking in Petri dishes at one end; a fine colony-counting tip is located at the other end.
  • Sterilized by radiation
  • Double use loops
  • 10μL
  • Ergonomic gripping from hexagonal design
  • Calibrated loops
Specifications
Sterility Sterile
Product Type Inoculating Loop