Learn More
- Home
- Products
- Specialty Lab Equipment, Instruments, and Apparatuses
- Crucibles
- Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Thermo Scientific Chemicals Al-23 Crucible, Tammann;Height x OD x ID (mm), 100 x 12 x 8;Volume (ml), 5;Tammann
Product Code. p-4865449
Change view
Click to view available options
Capacity (Metric):
0.2 mL
1 mL
2.3 mL
5 mL
8 mL
17 mL
30 mL
90 mL
123 mL
180 mL
300 mL
490 mL
790 mL
1070 mL
Diameter (Metric) Outer:
3mm
5mm
6mm
8mm
10mm
15mm
20mm
28mm
38mm
47mm
58mm
Diameter (Metric) Inner:
30mm
50mm
80mm
100mm
190mm
280mm
390mm
Height (Metric):
6mm
8mm
10mm
12mm
15mm
20mm
24mm
25mm
34mm
44mm
55mm
68mm
Unit Size:
Each
14 product options available for selection
| Product Code. | Capacity (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner | Height (Metric) | unitSize |
|---|---|---|---|---|---|
| 15403545 | 0.2 mL | 3mm | 30mm | 6mm | Each |
| 15423545 | 2.3 mL | 6mm | 80mm | 10mm | Each |
| 15433545 | 5 mL | 8mm | 100mm | 12mm | Each |
| 15443545 | 8 mL | 10mm | 100mm | 15mm | Each |
| 15413545 | 1 mL | 5mm | 50mm | 8mm | Each |
| 15463545 | 17 mL | 15mm | 100mm | 20mm | Each |
| 15483545 | 90 mL | 20mm | 280mm | 24mm | Each |
| 15493545 | 30 mL | 20mm | 100mm | 25mm | Each |
| 15443555 | 790 mL | 58mm | 280mm | 68mm | Each |
| 15403555 | 123 mL | 28mm | 190mm | 34mm | Each |
| 15423555 | 300 mL | 38mm | 280mm | 44mm | Each |
| 15413555 | 180 mL | 28mm | 280mm | 34mm | Each |
| 15433555 | 490 mL | 47mm | 280mm | 55mm | Each |
| 15453555 | 1070 mL | 58mm | 390mm | 68mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
This item is not returnable.
View return policy
Product Code.
15403545
Supplier
Thermo Scientific Chemicals
Supplier No.
033052.KF
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Diameter (Metric) Inner | 30mm |
| Material | Al-23 |
| Quantity | 1 Piece |
| Capacity (Metric) | 0.2 mL |
| Diameter (Metric) Outer | 3mm |
| Height (Metric) | 6mm |
| Product Type | Tammann Al-23 Crucibles |