Learn More
Thermo Scientific Chemicals Glassy Carbon Boat;Vol (ml), 3;Length (mm), 53;Width (mm), 16;Ht (mm),10
Product Code. p-6685042
Change view
Click to view available options
Dimensions (L x W x H):
29 x 16 x 10 mm
53 x 16 x 10 mm
76 x 33 x 25 mm
103 x 16 x 10 mm
Capacity (Metric):
2 mL
3 mL
10 mL
40 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Capacity (Metric) unitSize
15406295 29 x 16 x 10 mm 2 mL Each
15416295 53 x 16 x 10 mm 3 mL Each
15426295 103 x 16 x 10 mm 10 mL Each
15486345 76 x 33 x 25 mm 40 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15406295 Supplier Thermo Scientific Chemicals Supplier No. 039026.KT
Request Bulk or Custom Format
Offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Resistance to all wet decomposition agents and abrasive wear
  • No memory effects or contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistance to thermal shock allows rapid heating and cooling times
  • Good electrical conductivity
  • Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal­ growing such as doped halogenide crystals
TRUSTED_SUSTAINABILITY
Specifications
Type Glassy Carbon Boat
Capacity (Metric) 2 mL
Dimensions (L x W x H) 29 x 16 x 10 mm
Height (Metric) 10 mm
Length (Metric) 29 mm
Width (Metric) 16 mm