Learn More
Aldrin, TRC
Specifications
Specifications
| Chemical Name or Material | Aldrin |
| CAS | 309-00-2 |
| Molecular Formula | C12 H8 Cl6 |
| Quantity | 25 mg |
| Synonym | 1,4:5,8-Dimethanonaphthalene, 1,2,3,4,10,10-hexachloro-1,4,4a,5,8,8a-hexahydro-, (1α,4α,4aβ,5α,8α,8aβ)-, 1,4:5,8-Dimethanonaphthalene, 1,2,3,4,10,10-hexachloro-1,4,4a,5,8,8a-hexahydro-, endo,exo- (8CI), rel-(1R,4S,4aS,5S,8R,8aR)-1,2,3,4,10,10-Hexachloro-1,4,4a,5,8,8a-hexahydro-1,4:5,8-dimethanonaphthalene, 1,2,3,4,10,10-Hexachloro-1,4,4a,5,8,8a-hexahydro-1,4-endo-exo-5,8-dimethanonaphthalene, 1,2,3,4,10,10-Hexachloro-1,4,4a,5,8,8a-hexahydro-endo-1,4-exo-5,8-dimethanonaphthalene, Aldocit, Aldrex 25, Aldrex 40, Aldrin, Aldrine, Andrex, Andrex 40, Compound 118, ENT 15,949, HHDN, Kortofin, Octalene, SD 2794, Seedrin, Tatuzinho |
| InChI Formula | InChI=1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6+,7-,10+,11- |
| SMILES | ClC1=C(Cl)[C@@]2(Cl)[C@H]3[C@H]4C[C@H](C=C4)[C@H]3[C@]1(Cl)C2(Cl)Cl |
| IUPAC Name | (1R,4S,4aS,5S,8R,8aR)-rel-1,2,3,4,10,10-Hexachloro-1,4,4a,5,8,8a-hexahydro-1,4:5,8-dimethanonaphthalene |
| Molecular Weight (g/mol) | 364.91 |
| Formula Weight | 361.8757 |