Learn More
Novus Biologicals™ Albumin Native Protein
Product Code. 18462841 Shop All Bio Techne Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 18462841
Brand: Novus Biologicals™ NBP1970495mg
Native Protein
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P02768
For Use With (Application) Dot Blot, PAGE
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene ID (Entrez) 213
Quantity 5 mg
Storage Requirements Store at 4°C.
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbol ALB
Reconstitution Reconstitute with 500 μL of deionzied water.
Inhibitor Properties This product has been tested in SDS-Page and can be used in SDS, Western Blotting, ELISA experiments and other immunological assays.
Research Category Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Purity or Quality Grade Multi-step
Protein Albumin