Learn More
Emma Safety Footwear Alaska XD Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
| Design | Alaska XD |
| Material | Oiled Nubuck |
| Size | 37 |