Learn More
Alagebrium Chloride, TRC
Product Code. 30317293
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317293 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317293 Supplier Toronto Research Chemicals Supplier No. TRCA4704051G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Alagebrium Chloride
CAS 341028-37-3
Molecular Formula C13H14ClNOS
Quantity 1 g
Synonym 4,5-Dimethyl-3-(2-oxo-2-phenylethyl)thiazol-3-ium Chloride, ALT 711
InChI Formula InChI=1S/C13H14NOS.ClH/c1-10-11(2)16-9-14(10)8-13(15)12-6-4-3-5-7-12;/h3-7,9H,8H2,1-2H3;1H/q+1;/p-1
SMILES CC1=C(C)SC=[N+]1CC(=O)C2=CC=CC=C2.[Cl-]
IUPAC Name 2-(4,5-dimethyl-1,3-thiazol-3-ium-3-yl)-1-phenylethanone;chloride
Molecular Weight (g/mol) 267.77
Formula Weight 267.0485