missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ [Ala11,D-Leu15]-Orexin B
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
[Ala11,D-Leu15]-Orexin B is a highly potent and selective OX2 receptor agonist; displays 400-fold selectivity over OX1 receptors. EC50 values are 0.13 and 52 nM for human OX2 and OX1 receptors respectively.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Description | [Ala11,D-Leu15]-Orexin B |
| Molecular Weight (g/mol) | 2857.28 |
| Product Type | [Ala11,D-Leu15]-Orexin B |
| Quantity | 1 mg |
| Sequence | Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Ala-Gln-Arg-Leu-D-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction