Learn More
Al-23 Tube, Both ends open (Thin walls), O.D.: 7mm, I.D.: 6mm, Thermo Scientific™
Product Code. 11358247
Click to view available options
Quantity:
100 mm
200 mm
300 mm
Unit Size:
100mm
200mm
300mm
Product Code. 11358247
Brand: Thermo Scientific Alfa Aesar 031707.DM
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open (Thin walls)
Quantity 100 mm