Learn More
Al-23 Multibore Round Tube, O.D.: 2mm, Six 0.4mm Bores, Thermo Scientific™
Product Code. 11398507
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
Unit Size:
100mm
200mm
250mm
300mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398507 200 mm 200mm
11378507 100 mm 100mm
11318517 250 mm 250mm
11308517 300 mm 300mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11398507 Supplier Thermo Scientific Alfa Aesar Supplier No. 032601.DR
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Round Tube
Name Note Six bores
Quantity 200 mm