Learn More
Hamilton AirShield Tubing
Product Code. 31257789
Change view
Click to view available options
Length (English):
9.8 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English)
31257789 9.8 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31257789 Supplier Hamilton Supplier No. 9493901
 Restricted Item
Estimated Shipment: 07-12-2026
AirShield tubing designed for use with compatible systems. Supports efficient airflow and reliable operation in laboratory setups.
  • Designed for demanding laboratory environments requiring enhanced protection
  • Snorkel connects directly to the AirShield to supply clean external air
  • Delivers clean air from outside the immediate workspace
  • Creates a protective airflow around critical instrument components
  • Helps reduce exposure to contaminants and improve system reliability
Specifications
Product Type AirShield Tubing Kit
Length (English) 9.8 ft.