Learn More
Aalborg™ Air Direct Glass Flow Tube 150mm Reference Scale
Click to view available options
Airflow Rate:
1.25 L/min.
10 L/min.
100 mL/min.
1093 mL/min.
143 mL/min.
1637 mL/min.
19 mL/min.
2.5 L/min.
200 mL/min.
23 L/min.
25 mL/min.
264 mL/min.
30 L/min.
300 mL/min.
374 mL/min.
42 L/min.
49 mL/min.
50 mL/min.
500 mL/min.
5188 mL/min.
70 mL/min.
75 mL/min.
8505 mL/min.
Unit Size:
Each
Description
- Direct reading tubes are straightforward.
- The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
- Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
- Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Specifications
| Type | Air Direct Flow Tube |
| For Use With (Application) | For various fluids, gases or liquids, thanks to the correlation tables |
| Width (Metric) | 150 mm |
| Airflow Rate | 30 L/min. |