Learn More
Jenway™ Air Compressor 535 001
Product Code. 11405069
Change view
Click to view available options
Voltage:
230V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11405069 230V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11405069 Supplier Jenway™ Supplier No. 1265598
  • ONLINE EXCLUSIVE PRICE
    941.00€
    767.26€ / Pack of 1
    Save 173.74€
    18% Off
View all promotions
Estimated Shipment: 18-12-2026
Suitable air compressor to run a Jenway™ flame photometer. Jenway™ Air Compressor 535 001 is one of several accessories and consumables needed to complete a Jenway™ flame photometer assembly.
  • For use with Jenway™ Flame Photometer.
  • 220V/50Hz.
Specifications
Voltage 230V
Frequency 50Hz
Type Air Compressor