Learn More
Systec™ Air Compressor, Silent 50-N
Product Code. 15786482
Change view
Click to view available options
For Use With (Equipment):
Systec Autoclaves up to 50 L Chamber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15786482 Systec Autoclaves up to 50 L Chamber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15786482 Supplier Systec™ Supplier No. 8506
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Systec™ Air Compressor, Silent 50-N is designed to support cooling phase air pressure for Systec™ V, D, and HX series autoclaves with chamber volumes up to 50L.
  • Quiet operation
  • Supports air pressure in the cooling phase
  • Suitable for use with the following autoclaves:
    • V-55 to V-150
    • D-23 to D-200
    • HX-65 to HX-200
Specifications
Description Noise level: 45dB
For Use With (Equipment) Systec Autoclaves up to 50 L Chamber