Learn More
BTX™ AgilePulse™ MAX Safety Stand for Flatpack
Click to view available options
Connection Type:
BNC Cable
Unit Size:
Each
Description
- Cuvette module plugs directly into generator
- Holds one cuvette
- Safety swing shield secures cuvette in holder
Specifications
Specifications
| For Use With (Application) | Electroporation |
| For Use With (Equipment) | AgilePulse™ MAX, ECM™ 2001+ |
| Product Type | Safety Stands |
| Voltage | 500 to 1000 V |
| Description | AgilePulse MAX Safety Stand for Flatpack |
| Display | Touchscreen |
| Includes | Safety Stand Only |
| Sterility | Non-sterile |
| Connection Type | BNC Cable |
| Interface | Digital User |
| Show More |