Learn More
Simport™ Aggressive Adhesive Polyester Label Cable Ties
Product Code. 15216719 Shop All Brady Products
Change view
Click to view available options
Length (Metric):
201 mm
Unit Size:
Pack of 1500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15216719 201 mm Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15216719 Supplier Simport™ Supplier No. 134078
  • ONLINE EXCLUSIVE PRICE
    91.85€
    77.73€ / Pack of 1500
    Save 14.12€
    15% Off
 Restricted Item
In Stock
Aggressive Adhesive Polyester Label Cable Ties for inspection tagging.
  • Treated with epoxy resin coating.
  • Stainless steel grade 316.
  • Laser-engravable.
Specifications
Type Cable Tie
Material Stainless Steel (316)
Alternate Part Number SSTIEC-201-7316
Description Stainless Steel Coated Cable Tie
Color Silver
Length (Metric) 201 mm
Length (English) 7.9 in.
Width (Metric) 4.6 mm
Width (English) 0.181 in.
Dimensions (L x W) 201 x 4.6 mm
Thickness 0.5 mm
Quantity 100 Pk.
Certifications/Compliance RoHS Compliant
For Use With (Application) Inspection Tagging