Learn More
Kinematica Dispersing Aggregate Dia. 20mm PT-DA 20/2XEC-E116
Brand:  Kinematica 11030255
Dispersing Aggregate Dia. 20mm PT-DA 20/2XEC-E116
Product Code. 11974909
  • ONLINE EXCLUSIVE PRICE
    2247.00€
    2081.00€ / Each
    Save 166.00€
    7% Off
Only null left
Estimated Shipment: 29-12-2025