Learn More
Giese™ Agate Mortar with Pestle
Product Code. 10009520
Click to view available options
Capacity (Metric):
6 mL
8 mL
10 mL
13 mL
15 mL
24 mL
30 mL
50 mL
80 mL
130 mL
200 mL
350 mL
Unit Size:
Each
Product Code. 10009520
Brand: Giese™ 600M060
Grind up solid chemicals and crush solids into smaller pieces with this agate mortar and pestle. Giese™ Agate Mortar with Pestle features a smooth, dense, and hard surface.
  • Polished agate construction
  • For grinding and crushing
  • Durable
  • Available in different sizes
Specifications
Capacity (Metric) 13 mL
Diameter (Metric) Outer 60 mm
Product Type Mortar and Pestle
Material Agate Stone