missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose UHR

Product Code. 30017991
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30017991 500 g 500g
30017990 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017991 Supplier Melford Supplier No. A20135500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For DNA/RNA fragments 5-500bp.

For DNA/RNA fragments 5-500bp. Ideal for primer size. DNAse and RNase free at 3% concentration, Agarose Ultra High Resolution (UHR) presents a resolution of DNA fragments similar to gels made with polyacrylamide at concentrations of 8%. Gels with Agarose UHR are easier to prepare and are absolutely free of neurotoxic dangers inherent to polyacrylamide. While Agarose UHR can be dissolved by careful adherence to microwaving procedures, Agarose UHR gels are best prepared by autoclaving.

Specifications

Chemical Name or Material Agarose UHR
CAS 9012-36-6
Quantity 500 g
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.