Learn More
Xylem AG Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)
Product Code. p-200113486
Click to view available options
Refractive Index:
1.33659
1.34401
1.34782
1.34968
1.35171
1.35568
Unit Size:
25mL
Product Code. 30484478
Brand: Xylem 90501
Calibration fluid for calibration of refractometers.
  • Calibration Fluid
  • For Refractometers
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Fluid
Refractive Index 1.33659
For Use With (Equipment) Refractometer