Learn More
Xylem AG Refractometer Calibration Fluid 20 x 5 mL (UKAS/NIST)
Click to view available options
Refractive Index:
1.33659
1.34026
1.34401
1.34782
1.34968
1.35093
1.35171
1.35568
1.39986
Unit Size:
100mL
Description
- Calibration Fluid
- For Refractometers
Specifications
Specifications
| Type | Calibration Fluid |
| Refractive Index | 1.34782 |
| For Use With (Equipment) | Refractometer |