Learn More
Xylem AG Multi Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)
Click to view available options
For Use With (Application):
Beverage (USA)
Juice
Petrochemical
Pharma and Chem
Soda
Unit Size:
25mL
Description
- Calibration Fluid
- For Refractometers
Specifications
Specifications
| Type | Calibration Fluid |
| Refractive Index | Mixed |
| For Use With (Equipment) | Refractometer |
| For Use With (Application) | Juice |