Learn More
Macherey-Nagel™ Ag-Fix Test Strips
Brand: Macherey-Nagel™ 90741
This item is not returnable.
View return policy
Specifications
| Ochreous to Brown/Yellow to Blue | |
| Determination of silver and pH | |
| Silver and pH | |
| Silver, pH |
| Strip | |
| 0.5, 1, 2, 3, 5, 7, 10g/L Ag+ | |
| 100 Pack | |
| Test Strip |