Learn More
Macherey-Nagel™ Ag-Fix Test Strips
Brand:  Macherey-Nagel™ 90741
Product Code. 11366534
  • 34.30€ / Pack of 100
Specifications
Ochreous to Brown/Yellow to Blue
Determination of silver and pH
Silver and pH
Silver, pH
Strip
0.5, 1, 2, 3, 5, 7, 10g/L Ag+
100 Pack
Test Strip