missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies AG-490
Description
- AG-490 is a member of the tyrphostin family of small molecule inhibitors of tyrosine kinases including EGFR (Gazit et al.), HER2 (Gazit et al.), JAK2 (Burger et al.), JAK3 (Brown et al.), and STAT5a/b (Wang et al.) with IC50 values of 0.1, 13.5, 11 and 12 μM, respectively
- AG-490 does not inhibit LCK, LYN, BTK, SYK, or SRC tyrosine kinases (Meydan et al.)
Specifications
Specifications
| CAS | 133550-30-8 |
| Synonym | Tyrphostin AG-490 |
| Quantity | 10 mg |
| Molecular Formula | C17H14N2O3 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?