missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies AG-490

Product Code. 17190142
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 17190142

Brand: STEMCELL Technologies 72932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits EGFR, HER2, JAK2, JAK3, and STAT5a/b

  • AG-490 is a member of the tyrphostin family of small molecule inhibitors of tyrosine kinases including EGFR (Gazit et al.), HER2 (Gazit et al.), JAK2 (Burger et al.), JAK3 (Brown et al.), and STAT5a/b (Wang et al.) with IC50 values of 0.1, 13.5, 11 and 12 μM, respectively
  • AG-490 does not inhibit LCK, LYN, BTK, SYK, or SRC tyrosine kinases (Meydan et al.)

Specifications

CAS 133550-30-8
Synonym Tyrphostin AG-490
Quantity 10 mg
Molecular Formula C17H14N2O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.