missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1-d3, TRC

Product Code. 30323998
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323998 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323998 Supplier Toronto Research Chemicals Supplier No. TRCA3574621MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aflatoxin B1 O-methyl-d3
CAS 1217702-31-2
Molecular Formula C17 2H3 H9 O6
Quantity 1 mg
Synonym Aflatoxin B1-D3 (O-methyl-D3), Aflatoxin B1-d3, (6aR,9aS)-2,3,6a,9a-Tetrahydro-4-(methoxy-d3)-1H,11H-cyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-1,11-dione, Aflatoxin B1 D3 (methyl D3)
InChI Formula InChI=1S/C17H12O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8,17H,2-3H2,1H3/t8-,17+/m0/s1/i1D3
SMILES [2H]C([2H])([2H])Oc1cc2O[C@H]3OC=C[C@H]3c2c4OC(=O)C5=C(CCC5=O)c14
IUPAC Name (3S,7R)-11-(trideuteriomethoxy)-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
Molecular Weight (g/mol) 315.29
Formula Weight 315.0822
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.