Learn More
Aflatoxicol, TRC
Specifications
Specifications
| Chemical Name or Material | Aflatoxicol |
| CAS | 29611-03-8 |
| Molecular Formula | C17 H14 O6 |
| Quantity | 2.5 mg |
| Synonym | (1S,6aR,9aS)-2,3,6a,9a-Tetrahydro-1-hydroxy-4-methoxycyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-11(1H)-one, Aflatoxin R0, [1S-(1α,6aβ,9aβ)]-2,3,6a,9a-Tetrahydro-1-hydroxy-4-methoxy-cyclopenta[c]furo[3',2':4,5]furo[2,3-h][1]benzopyran-11(1H)-one |
| InChI Formula | InChI=1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8-9,17-18H,2-3H2,1H3/t8-,9-,17+/m0/s1 |
| SMILES | COc1cc2O[C@H]3OC=C[C@H]3c2c4OC(=O)C5=C(CC[C@@H]5O)c14 |
| IUPAC Name | (3S,7R,16S)-16-hydroxy-11-methoxy-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1,4,9,11,13(17)-pentaen-18-one |
| Molecular Weight (g/mol) | 314.29 |
| Formula Weight | 314.079 |