Learn More
Greiner Bio-One Advanced TC treated 6-well Cell Culture Microplates
GREENER_CHOICE
Product Code. 11920657
Click to view available options
Culture Area:
9.6 cm2
Unit Size:
Pack of 100
Product Code. 11920657
Brand: Greiner Bio-One 657960
Only null left
Add to basket
Add to basket
For high-throughput screening in cell based test systems.
  • Non-pyrogenic, non-cytotoxic
  • Improved cell adherence
  • Consistent cell attachment
  • Homogenous cell growth
  • In-vivo like morphology
  • Increased cell yield
  • Permits usage of serum-reduced or serum free media
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 5 mL (Well)
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Tissue Culture
Type Cell Culture Microplate
Volume (Metric) Working 2 to 5 mL
Well Count 6
Dnase Free Yes
Includes Lid
No. per Case 100
No. per Pack 1
RNase Free Yes
Dimensions (L x W x H) 128 x 85 x 19 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm