Learn More
HUBER Adjustable Bypass
Product Code. 15276557
Click to view available options
Size:
M16 x 1
M24 x 1.5
M30 x 1.5
M38 x 1.5
Unit Size:
Each
Product Code. 15276557
Brand: HUBER 9889
Specifications
For Use With (Equipment) Unistat
Temperature Range -20°C to +140°C
Type Bypass
Includes Manometer is not included.
Size M16 x 1