Learn More
3M™ Adflo™ Battery Charger (Li-ion)
Product Code. 15790008
Change view
Click to view available options
:
Battery Type: Lithium ion
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15790008 Battery Type: Lithium ion
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15790008 Supplier 3M™ Supplier No. 7000144537
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
3M™ Adflo™ Battery Charger (Li-ion) is a replacement charger suitable for use with Adflo™ Powered Air Respirator.
  • Replacement battery charger for Adflo Respirator Li-ion battery
  • Will charge Adflo standard battery in 4.5 hours or Adflo heavy-duty battery in 5.5 hours
  • For use with the 3M™ Adflo™ Powered Air Respirator
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion