missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Adenosine 5'-(a,b-methylene)diphosphate disodium salt
Description
Adenosine 5'-(α,β-methylene)diphosphate sodium salt is an ecto-5'-nucleotidase/CD73 inhibitor. Blocks ecto-5'-nucleotidase-mediated adenosine production by preventing the conversion of AMP to adenosine. Reverses doxorubicin -resistance in ecto-5'-nucleotidase-positive MDR cells (MCF7/A6 and L1210/A). Also increases rhodamine uptake and inhibits rhodamine efflux from ecto-5'-nucleotidase-positive MDR cells. Vasoconstrictor, induces inflammation in a rodent model of pleurisy, and reduces memory loss induced by Aβ1-42 in a mouse model of Alzheimer's disease.
Specifications
Specifications
| Chemical Name or Material | α,β-Methyleneadenosine 5'-diphosphate disodium salt |
| CAS | 104835-70-3 |
| Quantity | 10 mg |
| Target | 5'-Nucleotidase/CD73 |
| Molecular Formula | C11H15N5Na2O9P2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?