Learn More
Adenosine-2’,3’-cyclic Monophosphate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Adenosine-2',3'-cyclic Monophosphate Sodium Salt |
| CAS | 37063-35-7 |
| Molecular Formula | C10 H11 N5 O6 P . Na |
| Quantity | 25 mg |
| Synonym | Adenosine Cyclic 2',3'-(Hydrogen Phosphate) Monosodium Salt, 2',3'-Cyclic AMP Sodium Salt, Adenosine 2',3'-Cyclic Monophosphate Sodium Salt, Adenosine 2',3'-Cyclic Phosphate Sodium Salt, Cyclic Adenosine 2',3'-Monophosphate Sodium Salt, NSC 154034 |
| InChI Formula | InChI=1S/C10H12N5O6P.Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7-6(4(1-16)19-10)20-22(17,18)21-7;/h2-4,6-7,10,16H,1H2,(H,17,18)(H2,11,12,13);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1 |
| SMILES | [Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@H]4OP(=O)([O-])O[C@@H]34 |
| IUPAC Name | sodium;[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol |
| Molecular Weight (g/mol) | 351.188 |
| Formula Weight | 351.034 |