missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenine-2-d1, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Adenine-2-D |
| CAS | 109923-52-6 |
| Molecular Formula | C5 D H4 N5 |
| Quantity | 0.1 g |
| Synonym | 9 H-Purin-2-d-6-amine (ACI), 1 H-Purin-2-d-6-amine (9 CI), Adenine-2-d1, Adenine-2-D |
| InChI Formula | InChI=1 S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2 H,(H3,6,7,8,9,10)/i2D |
| SMILES | [2 H]c1nc(N)c2[nH]cnc2n1 |
| IUPAC Name | 2-deuterio-7 H-purin-6-amine |
| Molecular Weight (g/mol) | 136.1329 |
| Formula Weight | 136.0608 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction