Learn More
Miele Professional™ Additional Suction Lance PLW8636 APLW 887
Product Code. 30533217 Shop All Miele Products
Change view
Click to view available options
Product Type:
Filter
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
30533217 Filter 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533217 Supplier Miele Professional™ Supplier No. 12426920
Suction lance for external canisters for PLW 8636
  • Suction lance for external canisters for PLW 8636
  • For 10 - 20 L canisters, positioned outside the machine
  • The suction lance features level detection
  • Installed by Customer Service
TRUSTED_SUSTAINABILITY
Specifications
Product Type Filter
For Use With (Equipment) Washer-Disinfectors
Quantity 1 Ea.