Learn More
Memmert™ Additional Perforated Stainless Steel Shelf for UN55
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55 Oven
Unit Size:
Each
Description
- Perforated Stainless Steel Shelf
- Accessory for Memmert ovens and incubators
Specifications
Specifications
| For Use With (Equipment) | UN55 Oven |
| Type | Perforated Shelf |
| Material | Stainless Steel |