Learn More
Epredia™ Adcell™ PTFE Diagnostic Slides
Product Code. 17342650 Shop All Epredia Products
Click to view available options
Well Count:
3
12
Color:
Black
White
Unit Size:
Pack of 100
Product Code. 17342650
Brand: Epredia™ X5XER202WAD1
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
  • Binds and keeps liquids applied to the well area
  • PTFE Construction
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 25 mm
Width (Metric) 75 mm
Material PTFE
Thickness (Metric) 1 mm
Type ADCELL
Well Count 12
Color White
Diameter (Metric) 5 mm Well
Quantity 100/Pk.