missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Adavosertib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Adavosertib is a potent and selective Wee1 inhibitor (IC50 = 5.2 nM). In a panel of 223 serine/threonine or tyrosine kinases, Adavosertib is highly selective for Wee 1 over all except 8 kinases. In particular Adavosertib shows >100-fold selectivity over human Myt1. Adavosertib blocks CDC2Y15 phosphorylation and Gemcitabine (Cat. No. 3259)-induced DNA damage checkpoint, leading to premature mitotic entry and apoptosis in p53-deficient tumor cell lines. Adavosertib also potentiates the cytotoxic effects of Pemetrexed (Cat. No. 6185), Doxorubicin (Cat. No. 2252), Camptothecin (Cat. No. 1100) and Mitomycin C (Cat. No. 3258) in vitro. Adavosertib synergizes with gemcitabine to achieve tumor regressions, selectively in p53-deficient pancreatic cancer xenografts.
Specifications
Specifications
| Chemical Name or Material | 1,2-Dihydro-1-[6-(1-hydroxy-1-methylethyl)-2-pyridinyl]-6-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-2-(2-propen-1-yl)-3 H-pyrazolo[3,4-d]pyrimidin-3-one |
| CAS | 955365-80-7 |
| Quantity | 10 mg |
| Target | Checkpoint Control Kinase Inhibitors |
| Molecular Formula | C27H32N8O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction