Learn More
Lenz Laborglasintrumente Adapters Straight, for Distilling Receivers acc. to Bredt
Product Code. p-7222649
Click to view available options
Standard Taper Joint:
Upper socket NS 14/23, Lower socket NS 19/26
Upper socket NS 29/32, Lower socket NS 29/32
Unit Size:
Each
Product Code. 10602092
Brand: Lenz Laborglasintrumente 05285106
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Straight adapters
  • With two NS sockets
  • Vacuum connection: GL 18
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Standard Taper Joint Upper socket NS 14/23, Lower socket NS 19/26