Learn More
Fisherbrand™ Adapters for Sample Tubes
Product Code. 11833503
Change view
Click to view available options
For Use With (Equipment):
0.4mL sample tubes
0.5mL sample tubes
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11833503 0.4mL sample tubes Pack of 6
10732711 0.5mL sample tubes Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11833503 Supplier Fisherbrand™ Supplier No. C1206
  • ONLINE EXCLUSIVE PRICE
    49.35€
    45.12€ / Pack of 6
    Save 4.23€
    8% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
Excellent for use with micro centrifuges. Fisherbrand™ Adapters for Sample Tubes are durable and resilient.
  • White in color
  • Available for different tube sizes
Specifications
Type Adapter
For Use With (Equipment) 0.4mL sample tubes
No. per Pack 6/Pk.