Learn More
Carl Zeiss™ Adapters
Product Code. 10738683
Click to view available options
Type:
Adapter 60N - T2, 1.0X
DSLR P95-T2 1.6x Adapter
Preparation Piece 60-44
Video Adapter 44 C 2/3, 1,0X
Video Adapter 44 C, 0.66 in., 0.63X
Video Adapter 44, 0.5 in., 0.5X
Video Adapter 60 C, 0.5 in., 0.5X
Unit Size:
Each
Product Code. 10738683
Brand: Carl Zeiss™ 4561400000000