Learn More
asecos™ Adapter for UFA
Product Code. 11918827 Shop All asecos Products
Change view
Click to view available options
For Use With (Equipment):
For models before type VBF.197 and other brands (with exhaust air plug DN 75 mm)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11918827 For models before type VBF.197 and other brands (with exhaust air plug DN 75 mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11918827 Supplier asecos™ Supplier No. 15784
  • ONLINE EXCLUSIVE PRICE
    211.35€
    156.62€ / Pack of 1
    Save 54.73€
    25% Off
 Restricted Item
Estimated Shipment: 01-03-2027
Adapter for use with recirculating air filter system UFA
  • For model series before type VBF.197 and third-party cabinets (with connection DN 75 mm)
  • Height = 160 mm
Specifications
Description Adapter for recirculation air filter system
For Use With (Equipment) For models before type VBF.197 and other brands (with exhaust air plug DN 75 mm)
Surface Coating Paint
Weight (Metric) 2.50 kg
Material Steel
Product Type Adapter