Learn More
Epredia™ Adamas Blades for Disposable Magnetic Blade Carriers
Product Code. 12627826 Shop All Epredia Products
Click to view available options
Length (Metric):
60 mm
80 mm
Quantity:
10/Cs.
50/Cs.
Unit Size:
Pack of 10
Pack of 50
Product Code. 12627826
Brand: Epredia™ 160050
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Choose between two sizes of Epredia™ Adamas Blades for Disposable Magnetic Blade Carriers, available in 60 mm and 80 mm.
TRUSTED_SUSTAINABILITY
Specifications
Description 80 x 18mm; 50/cs.
Product Type Disposable, Magnetic
For Use With (Equipment) Disposable Magnetic Blade Carrier
Quantity 50/Cs.
Length (Metric) 80 mm
Width (Metric) 18 mm