Learn More
KD Scientific Adagio Syringe Pump Control Software
Product Code. 13433649
Change view
Click to view available options
For Use With (Equipment):
Legato Syringe Pumps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13433649 Legato Syringe Pumps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13433649 Supplier KD Scientific Supplier No. 788000
  • ONLINE EXCLUSIVE PRICE
    722.00€
    628.76€ / Pack of 1
    Save 93.24€
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-01-2027
For Legato™ Syringe Pumps. One license for software on USB key.
  • Low cost simple installation
  • Flow evolution graph
  • Import and Export programs
  • Quick and easy manual pump control
  • Monitor one or more pumps
  • Program data logging
Specifications
Type Control Software
For Use With (Equipment) Legato Syringe Pumps
Voltage 0/1 V
Product Title
Select an issue
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.