Learn More
Thermo Scientific Chemicals ADA, 98+%
Description
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼218°C (decomposition) |
| Quantity | 100 g |
| Beilstein | 1787181 |
| Merck Index | 14,147 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 190.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | ADA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.