missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACVRL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | ACVRL1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883544
|
Proteintech
14745-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16873544
|
Proteintech
14745-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
The TGF-beta superfamily of ligands exerts its biological activity by binding to heteromeric receptor complexes two types (I and II) of the serine/threonine kinases. ALK1, like the other type I receptors, contains a cysteine rich domain with conserved cysteine spacing in the extracellular region and a glycine and serine rich domain preceding the kinase domain. The reduced human ALK1 monomer has a calculated molecular mass of approximately 37.3 kDa. Due to glycoslyation, the recombit human protein migrates as a 50-55 kDa protein in SDS-PAGE under reducing conditions. Human and mouse ALK1 share approximately 71% amino acid sequence identity in their extracellular domains. ALK1 is highly expressed in endothelial cells and other vascular tissues.Specifications
ACVRL1 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P37023 | |
94 | |
ACVRL1 Fusion Protein Ag6449 | |
Primary | |
-20°C | |
ACVRL1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Activin receptor like kinase 1, ACVRL1, ACVRLK1, ALK 1, ALK1, HHT, HHT2, ORW2, SKR3, TSR I | |
ACVRL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |