Learn More
Socorex™ Acura™ Electro 936 Macropipette
Product Code. 11889223
Click to view available options
Volume (Metric):
0.25 to 5 mL
0.5 to 10 mL
Unit Size:
Each
Product Code. 11889223
Brand: Socorex™ 936.05
Ergonomically friendly, this electronic single-channel macropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 936 Macropipette charges quickly and has a long working autonomy.
  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Interchangeable nozzle protection filter.
  • Adapters for Pasteur pipette and straw tips.
  • Outstanding accuracy and precision.
Specifications
Accuracy <±1.2% - <±0.5%
Battery Type 4.8V NiMH
Certifications/Compliance EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
Number of Channels 1
Product Line Acura
Type Electronic Macropipette
Volume (Metric) 0.25 to 5 mL
Display Right/Left LCD
Includes QC certificate, Qualitips™ pipette tip samples and operating instructions.
Increments 0.05 mL
Precision <0.6% - <0.15%
Quantity 1 Each
Recharge Time <1.5 Hours
Warranty 24 Months